1,4-Dithioerythritol
Supplier: Serva
Certificates
About this item
Synonyms: Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
CAS Number:6892-68-8
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00063750
:154.25 g/mol
Boiling Pt:286 °C (1013 hPa)
Melting Pt:82…83 °C
Storage Temperature:Ambient
EINECS:229-998-8
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together