Order Entry
Belgium
ContactUsLinkComponent
1,4-Dithioerythritol
1,4-Dithioerythritol
Catalog # MOLEM62236573
Supplier:  Molekula
CAS Number:  
1,4-Dithioerythritol
Catalog # MOLEM62236573
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
 :
 :154.25 g/mol
Boiling Pt:286 °C (1013 hPa)
Melting Pt:82…83 °C
EINECS:229-998-8
 :GHS07
Storage Temperature:Ambient
CAS Number:6892-68-8
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00063750