Orders LinkContactUsLinkComponent
Pasteur pipettes
Pasteur pipettes
Supplier:   Brand
Pasteur pipettes
Supplier:  Brand
Certificates
Pasteur pipettes
Pasteur pipettes
2 Options Available Below
Product Details & Documents
Soda-lime-glass.
Long drawn-out capillary tip. Suction end with constriction for a cotton plug. int.Ø of the tip: 1,0 mm.
Certifications: ISO 7712
Documents
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Length
    Packed
    Availability
    Price
  • 2,0 ml
    7 mm
    145 mm
    250/box, 4 boxes/pack
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 80,70 €
    80,70 €
    1 * 1.000 ST
       80,70 €
  • 2,0 ml
    7 mm
    225 mm
    250/box, 4 boxes/pack
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 88,60 €
    88,60 €
    1 * 1.000 ST
       88,60 €