Orders LinkContactUsLinkComponent
Slide boxes, PS
Slide boxes, PS
Supplier:   Brand
Slide boxes, PS
Supplier:  Brand
Certificates
Slide boxes, PS
3 Options Available Below
Product Details & Documents
Grey polystyrene slide boxes.
Product Family Options
Product Information
  • Colour
    Material
    No. of slides
    W×D×H
    Availability
    Price
  • Grey
    PS
    25
    120×96×35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,60 €
    21,60 €
    1 * 1 ST
       21,60 €
  • Grey
    PS
    50
    230×97×35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,70 €
    31,70 €
    1 * 1 ST
       31,70 €
  • Grey
    PS
    100
    230×187×35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,90 €
    42,90 €
    1 * 1 ST
       42,90 €