Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Stopcocks, needle, Nalgene®

9,53 mm, 15,88 mm, NALGENE® HDPE tanks up to 378,541 L, 1-1/8" - 12 straight female thread
  NALG96423-0100
   Low Manufacturing Impact
Stopcocks, needle, Nalgene®
  NALG96423-0100
   Low Manufacturing Impact
Stopcocks, needle, Nalgene®

9,53 mm, 15,88 mm, NALGENE® HDPE tanks up to 378,541 L, 1-1/8" - 12 straight female thread

 
275,00 €
   
1 * 1 ST
 

Spigot is installed on a threaded HDPE boss which is welded onto the tank at the factory.

  • Boss fits only Nalgener tanks
  • Spigot Material: Polypropylene
  • Accepts ⁵/₈" inner Ø tubing
 
  • For:
    NALGENE® HDPE tanks up to 378,541 L
  • Hole Ø:
    9,53 mm
  • Thread:
    1-1/8" - 12 straight female thread
  • For tube I-Ø:
    15,88 mm
  • Cat. No.:
    NALG96423-0100
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact