Orders LinkContactUsLinkComponent
Desiccator lids, with socket
Desiccator lids, with socket
 :   Brand
Desiccator lids, with socket
 :  Brand
Desiccator lids, with socket
Desiccator lids, with socket
Dessicator lids with socket 24/29, made of borosilicate glass 3.3.
Product Family Options
  • Description
    Type
    Nominal size
    Height
    Ext.Ø
  • Desicator lid, Hxø: 84x215 mm
    With socket, Novus
    150 DN
    85 mm
    215 mm
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • Desicator lid, Hxø: 92x270 mm
    With socket, Novus
    200 DN
    94 mm
    270 mm
    1 * 1 ST
    1 * 1 ST - 176,00 €
    176,00 €
    1 * 1 ST
       176,00 €
  • Desicator lid, Hxø: 118x250 mm
    With socket, Novus
    250 DN
    109 mm
    320 mm
    1 * 1 ST
    1 * 1 ST - 198,00 €
    198,00 €
    1 * 1 ST
       198,00 €