Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Acetaldehyde

Glass bottle, 100 mL
Catalog # ROTH3004.1
Flammable
Health
Irritant
Marketsource
Supplier:   Roth Carl
Acetaldehyde
Catalog # ROTH3004.1
Supplier:  Roth Carl
Marketsource
CAS Number:  75-07-0
Acetaldehyde
Acetaldehyde

Glass bottle, 100 mL

CAS Number:  75-07-0
Price
54,90 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Ethanal
  • CAS Number:75-07-0
  • Formula:CH₃CHO
  • MDL Number:MFCD00006991
  • Molecular Weight:44.05 g/mol
  • Merck Index:12,00037
  • Boiling Pt:20.1 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.78 g/cm³ (20 °C)
  • Flash Pt:–27 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-836-8
  • UN:1089
  • ADR:3,I
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications