


Lanthanum(III) chloride heptahydrate analytical reagent, Supelco®
Catalog # 1.12219.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:91 °C Elimination of water of crystallisation
- Molar Mass:371.37 g/mol
- Bulk Density:900 kg/m3
- pH:5 (100 g/l, H2O, 25 °C)
Specification Test Results
About this item
:
Boiling Pt:∼860 °C (1013 hPa)
Melting Pt:96 °C
:M-Clarity™ quality level = MQ100
:371.37 g/mol
EINECS:233-237-5
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:10025-84-0
Formula:LaCl₃·7H₂O
MDL Number:MFCD00149756