Order Entry
Belgium
ContactUsLinkComponent
 
Magnesium chloride hexahydrate 99.0-101.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
  25108.295
 :  
undefined
Magnesium chloride hexahydrate 99.0-101.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
  25108.295
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay:
    99.0 - 101.0 %
  • Acidity or alkalinity:
    Passes test Ph.Eur.
  • Appearance of solution (20 %; water):
    Passes test
  • Identification B:
    Passes test Ph.Eur.
  • Identification C:
    Passes test Ph.Eur.
  • pH (20°C; 5 %):
    5.0 - 6.5
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Total N (Nitrogen):
    Max. 20 ppm
  • Water:
    51.0 - 55.0 %
  • Br (Bromide):
    Max. 0.05 %
  • NH4 (Ammonium):
    Max. 20 ppm
  • NO3 (Nitrate):
    Max. 10 ppm
  • PO4 (Phosphate):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • As (Arsenic):
    Max. 2 ppm
  • Ba (Barium):
    Max. 20 ppm
  • Ca (Calcium):
    Max. 30 ppm
  • Cu (Copper):
    Max. 5 ppm
  • Fe (Iron):
    Max. 5 ppm
  • K (Potassium):
    Max. 10 ppm
  • Mn (Manganese):
    Max. 5 ppm
  • Na (Sodium):
    Max. 10 ppm
  • Pb (Lead):
    Max. 5 ppm
  • Sr (Strontium):
    Max. 50 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

 

 

 :
 :203.3 g/mol
 :13,05684
 :1412 °C (1013 hPa)
 :117 °C
 :1.57 g/cm³ (25 °C)
 :Ambient
 :232-094-6
 :7791-18-6
 :MgCl₂·6H₂O
 :MFCD00149781