Order Entry
Belgium
ContactUsLinkComponent
 
Magnesium chloride hexahydrate 99-101%, Puriss., meets analytical specification of BP, FCC, Ph. Eur., E511 (max. 0.0001% Al), Fluka™
 
Magnesium chloride hexahydrate 99-101%, Puriss., meets analytical specification of BP, FCC, Ph. Eur., E511 (max. 0.0001% Al), Fluka™

 

  • Assay:
    99.0 - 101.0 %
  • Water (Karl Fischer):
    51.0 - 55.0 %
  • Free acid (as HCl):
    Max. 0.02 %
  • Free alkali (as MgO):
    Max. 0.01 %
  • Acidity or alkalinity:
    complying
  • Ammonium (NH₄):
    Max. 50 ppm
  • Aluminium (Al):
    Max. 1 ppm
  • Arsenic (As):
    Max. 2 ppm
  • Barium (Ba):
    Max. 0.002 %
  • Calcium (Ca):
    Max. 0.1 %
  • Iron (Fe):
    Max. 5 ppm
  • Mercury (Hg):
    Max. 1 ppm
  • Potassium (K):
    Max. 500 ppm
  • Sodium (Na):
    Max. 0.5 %
  • Lead (Pb):
    Max. 2 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Bromide (Br):
    Max. 500 ppm
  • Sulfate (SO₄):
    Max. 100 ppm
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying

 

 

Frequently Bought Together