ContactUsLinkComponent
Calcium chloride dihydrate ≥99% (by complexometric analysis), white solid, Molecular Biology Grade, Millipore®
  208291-250
 :  
Calcium chloride dihydrate ≥99% (by complexometric analysis), white solid, Molecular Biology Grade, Millipore®
  208291-250
 :  
  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545
 :
 :147.01 g/mol
 :13,01660
 :1600 °C (1013 hPa)
 :176 °C
 :1.85 g/cm³ (20 °C)
 :Ambient
 :233-140-8
 :GHS07
 :10035-04-8
 :CaCl₂·2H₂O
 :MFCD00149613