Order Entry
Belgium
ContactUsLinkComponent
Sodium hydrogen sulphate monohydrate, EMSURE® for analysis, Supelco®
Sodium hydrogen sulphate monohydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Sodium hydrogen sulphate monohydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    1080 g/l (20 °C)
  • Melting Point:
    183 °C (decomposition)
  • Molar Mass:
    138.07 g/mol
  • Bulk Density:
    900 - 970 kg/m3
  • Density:
    2.12 g/cm3 (20 °C)
  • pH:
    1 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together