Sodium hydrogen sulphate monohydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium bisulphate monohydrate
CAS Number:10034-88-5
:
Formula:NaHSO₄·H₂O
MDL Number:MFCD00149210
Density:2.1 g/cm³ (20 °C)
Molecular Weight:138.08 g/mol
Melting Pt:∼183 °C (dec.)
ADR:8 III
Merck Index:14,8586
EINECS:231-665-7
UN:3260
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together