Lithium sulphate monohydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.05694.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:340 g/l (20 °C)
- Melting Point:120 °C
- Molar Mass:127.96 g/mol
- Bulk Density:940 kg/m3
- Density:2.05 g/cm3 (20 °C)
- pH:4.5 - 6.0 (50 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:127.96 g/mol
Melting Pt:120
EINECS:233-820-4
:GHS07
Storage Temperature:Ambient
CAS Number:10102-25-7
Formula:Li₂SO₄·H₂O
MDL Number:MFCD00149766
Density:2.05 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300