Order Entry
Belgium
ContactUsLinkComponent
 
Sodium thiosulphate pentahydrate, Suprapur® for trace analysis, Supelco®
  1.06509.0100
 :  Merck
 :  
Sodium thiosulphate pentahydrate, Suprapur® for trace analysis, Supelco®
  1.06509.0100
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Solubility:
    701 g/l (20 °C)
  • Melting Point:
    48 °C
  • Molar Mass:
    248.21 g/mol
  • Bulk Density:
    1000 kg/m3
  • Density:
    1.74 g/cm3 (20 °C)
  • pH:
    6.0 - 7.5 (100 g/l, H2O, 20 °C)

 

 

 :
 :248.19 g/mol
 :12,08844
 :48 °C
 :1.67 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :Ambient
 :231-867-5
 :10102-17-7
 :Na₂S₂O₃.5H₂O
 :MFCD00149186