Order Entry
Belgium
ContactUsLinkComponent
Disodium hydrogen phosphate heptahydrate, EMSURE® ACS for analysis, Supelco®
Disodium hydrogen phosphate heptahydrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck
Certificates
Disodium hydrogen phosphate heptahydrate, EMSURE® ACS for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (acidimetric):
    98.0 - 102.0 %
  • Identity:
    passes test
  • Insoluble matter:
    ≤ 0.005 %
  • pH-value (5 %; water, 25 °C):
    8.7 - 9.3
  • Chloride (Cl):
    ≤ 0.001 %
  • Sulfate (SO₄):
    ≤ 0.005 %
  • Heavy metals (ACS):
    ≤ 0.001 %
  • Fe (Iron):
    ≤ 0.001 %

Specification Test Results

Related Information

Frequently Bought Together