Disodium hydrogen phosphate dihydrate ≥99%, GPR RECTAPUR®
28040.291
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 10 ppm
- Cl (Chloride):Max. 0.02 %
- SO4 (Sulphate):Max. 0.05 %
- As (Arsenic):Max. 2 ppm
- Fe (Iron):Max. 50 ppm
:
:177.99 g/mol
:92.5 °C
:2.1 g/cm³ (20 °C)
:Ambient
:231-448-7
:10028-24-7
:Na₂HPO₄·2H₂O
:MFCD00149182