Order Entry
Belgium
ContactUsLinkComponent
 
Disodium hydrogen phosphate dihydrate 98.0-101.0% (dry basis), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
New Product
 
Disodium hydrogen phosphate dihydrate 98.0-101.0% (dry basis), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
New Product
 

 

  • Assay (Calculated on Dried Substance According to Ph. Eur.):
    98.0 - 101.0%
  • Assay (Calculated on Dried Substance According to USP):
    98.0 - 100.5%
  • Identity:
    Passes Test
  • Appearance of Solution:
    Passes Test
  • In Water Insoluble Matter:
    ≤0.3%
  • pH-Value (5%; Water):
    8.9 - 9.2
  • Chloride (Cl):
    ≤0.005%
  • Sulfate (SO₄):
    ≤0.02%
  • Heavy Metals (as Pb):
    ≤0.001%
  • As (Arsenic):
    ≤0.0002%
  • Fe (Iron):
    ≤0.001%
  • Sodium Dihydrogen Phosphate (NaH₂PO₄):
    ≤1.7%
  • Residual Solvents (ICH Q3C):
    Excluded by Manufacturing Process
  • Substances Reducing Potassium Permanganate (as O):
    Passes Test
  • Loss on drying (130 °C):
    19.5 - 21.0 %

 

Frequently Bought Together