Order Entry
Belgium
ContactUsLinkComponent
 
Disodium hydrogen phosphate dihydrate 99+% for HPLC
 
undefined
Disodium hydrogen phosphate dihydrate 99+% for HPLC

 

  • Appearance:
    White Crystalline powder or crystals
  • UV:
    (0.5 M in water)
  • pH:
    7.5 to 9.2 (5 % at 25°C)
  • Titration with HCl:
    ≥99.0 %
  • Chloride (Cl):
    ≤50 ppm
  • Lead (Pb):
    ≤5 ppm

 

 

Frequently Bought Together