Disodium hydrogen phosphate dodecahydrate 98.5-102.5% (by acidimetry), crystals, EMSURE® Ph. Eur., BP, JP, USP, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10039-32-4
:
Formula:Na₂HPO₄·12H₂O
MDL Number:MFCD00149181
:358.14 g/mol
Merck Index:13,08733
Melting Pt:35 °C
Density:1.52 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:231-448-7
Specification Test Results
- Assay (alkalimetric):99.0 - 102.0 %
- Identity:passes test
- Appearance of solution:passes test
- Sodium Dihydrogene phospate:≤ 2.5 %
- pH-value (5 %; water):9.0 - 9.3
- Chloride (Cl):≤ 0.0005 %
- Sulfate (SO₄):≤ 0.005 %
- Total nitrogen (N):≤ 0.001 %
- Heavy metals (as Pb):≤ 0.0005 %
- As (Arsenic):≤ 0.00005 %
- Cu (Copper):≤ 0.0002 %
- Fe (Iron):≤ 0.0005 %
- K (Potassium):≤ 0.005 %
- Pb (Lead):≤ 0.0005 %
- Reducing substances:passes test
- Loss on drying (130 °C):57.0 - 61.0 %
- Corresponds to ISO,Reag. Ph Eur:passes test
- Water (according to Karl Fischer):57.0 - 61.0 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06579.0500_TDS_EN MERC1.06579.1000_TDS_EN ProductPage_106579 ProductPage_106579
Product specification
Frequently Bought Together