
Trisodium phosphate dodecahydrate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10101-89-0
:
Formula:Na₃PO₄·12H₂O
MDL Number:MFCD00149198
:380.12 g/mol
Merck Index:12,08808
Melting Pt:∼75 °C
Density:1.62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-509-8
UN:3262
ADR:8,III
:GHS07
Specification Test Results
- Assay:Min. 99 %
- Identification:Passes test
- Free alkali:Max. 0.7 meq/g
- Total N (Nitrogen):Max. 100 ppm
- Cl (Chloride):Max. 25 ppm
- SO4 (Sulphate):Max. 100 ppm
- As (Arsenic):Max. 1 ppm
- Cu (Copper):Max. 25 ppm
- Fe (Iron):Max. 10 ppm
- Pb (Lead):Max. 10 ppm
Specification Test Results
Frequently Bought Together