Order Entry
Belgium
ContactUsLinkComponent
Trisodium phosphate dodecahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Irritant
Trisodium phosphate dodecahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Irritant
Trisodium phosphate dodecahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    285 g/l (20 °C)
  • Melting Point:
    75 °C Elimination of water of crystallisation
  • Molar Mass:
    380.18 g/mol
  • Bulk Density:
    950 kg/m3
  • Density:
    1.62 g/cm3 (20 °C)
  • pH:
    12 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together