
Trisodium phosphate dodecahydrate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10101-89-0
:
Formula:Na₃PO₄·12H₂O
MDL Number:MFCD00149198
:380.12 g/mol
Merck Index:14,8662
Melting Pt:∼75 °C (dec.)
Density:1.62
Storage Temperature:Ambient
EINECS:231-509-8
ADR:8,III
REACH:01-2119489800-32-xxxx
:GHS05
Related Information
Frequently Bought Together