Order Entry
Belgium
ContactUsLinkComponent
Tetrasodium diphosphate decahydrate ≥97%, GPR RECTAPUR®
Irritant
Tetrasodium diphosphate decahydrate ≥97%, GPR RECTAPUR®
Catalog # 28053.295
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Tetrasodium diphosphate decahydrate ≥97%, GPR RECTAPUR®
Catalog # 28053.295
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 97 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 0.05 %
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :446.06 g/mol
Melting Pt:990 °C
Density:1.82 g/cm³ (25 °C)
EINECS:231-767-1
 :GHS07
Storage Temperature:Ambient
CAS Number:13472-36-1
Formula:Na₄P₂O₇·10H₂O
MDL Number:MFCD00149200