Sodium molybdate(VI) dihydrate ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10102-40-6
:
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170
:241.95 g/mol
Merck Index:13,08718
Melting Pt:687 °C
Density:3.28 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-551-7
Specification Test Results
- Assay:Min. 99.0 %
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 50 ppm
- NH4 (Ammonium):Max. 20 ppm
- SO4 (Sulphate):Max. 0.05 %
- Fe (Iron):Max. 20 ppm
Specification Test Results
Related Information
Frequently Bought Together