Sodium molybdate(VI) dihydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10102-40-6
:
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170
:241.95 g/mol
Merck Index:13,08718
Melting Pt:687 °C
Density:3.28 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:231-551-7
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay (precipitative titration):≥ 99.5 %
- pH-value (5 %; water, 25 °C):7.0 - 10.5
- Insoluble matter:≤ 0.005 %
- Chloride (Cl):≤ 0.005 %
- Nitrate (NO₃):≤ 0.005 %
- Phosphate (PO₄):≤ 0.0005 %
- Phosphate, Arsenate, Silicate (as PO₄):≤ 0.001 %
- Sulfate (SO₄):≤ 0.005 %
- Heavy metals (as Pb):≤ 0.0005 %
- Fe (Iron):≤ 0.001 %
- NH₄ (Ammonium):≤ 0.001 %
- Pb (Lead):≤ 0.001 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06521.0100_TDS_EN ProductPage_106521
Product specification
msds_106521
SDS
Frequently Bought Together