

Sodium perchlorate monohydraté, EMSURE® pour analyses, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7791-07-3
:
Formula:NaClO₄·H₂O
MDL Number:MFCD00149164
:140.46 g/mol
Merck Index:12,08798
Melting Pt:130 °C
Density:2,02 g/cm³ (25 °C)
Storage Temperature:Température ambiante
EINECS:231-511-9
ADR:5.1,II
:M-Clarity™ (Niveau de qualité) = MQ300
UN:1502
:GHS03
:GHS07
Specification Test Results
- Assay (alkalimetric):99.0 - 102.0 %
- pH-value (5 %; water):4.5 - 7.0
- Chloride (Cl):≤ 0.002 %
- Chloride, Chlorate (as Cl):≤ 0.002 %
- Sulfate (SO₄):≤ 0.002 %
- Total nitrogen (N):≤ 0.0005 %
- Ca (Calcium):≤ 0.002 %
- Fe (Iron):≤ 0.0003 %
- K (Potassium):≤ 0.005 %
- Heavy metals (as Pb):≤ 0.0005 %
Specification Test Results
Related Information
Frequently Bought Together