Order Entry
Belgium
ContactUsLinkComponent
Sodium perchlorate monohydrate ≥99.0%, HiPerSolv CHROMANORM® for HPLC
 
 
Sodium perchlorate monohydrate ≥99.0%, HiPerSolv CHROMANORM® for HPLC
  153233M
 :  
undefined
 
 
Sodium perchlorate monohydrate ≥99.0%, HiPerSolv CHROMANORM® for HPLC
  153233M
 :  

 

  • Pack type:
    Metal can
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Assay (argentometric, as monohydrate):
    Min. 99.0 %
  • Transmittance (254 nm) (1 mol/l):
    Min. 96 %
  • Chloride, chlorate (as Cl):
    Max. 0.002 %
  • SO4 (Sulphate):
    Max. 0.002 %
  • Ca (Calcium):
    Max. 0.002 %
  • Fe (Iron):
    Max. 0.0003 %
  • K (Potassium):
    Max. 0.005 %
  • Pb (Lead):
    Max. 0.0005 %

 

 

 :
 :140.46 g/mol
 :12,08798
 :130 °C
 :2.02 g/cm³ (25 °C)
 :Ambient
 :231-511-9
 :1502
 :5.1,II
 :GHS03
 :GHS07
 :7791-07-3
 :NaClO₄·H₂O
 :MFCD00149164