di-Sodium tetraborate decahydrate, EMPROVE® ESSENTIAL JP, NF, BP, Ph. Eur. suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
CAS Number:1303-96-4
:
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193
:381.37 g/mol
Merck Index:13,08662
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1.73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together