Order Entry
Belgium
ContactUsLinkComponent
Orthophosphoric acid ≥85.0-88.0% NF, Multi-Compendial, J.T.Baker®
Corrosive
Irritant
Orthophosphoric acid ≥85.0-88.0% NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Orthophosphoric acid ≥85.0-88.0% NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    Passes Test
  • Meets E.P. Chemical Specifications:
    Passes Test
  • Meets B.P. Chemical Specifications:
    Passes Test
  • Meets N.F. Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    ≤0.001%
  • NF - Identification:
    Passes Test
  • NF - Nitrate (NO₃):
    Passes Test
  • NF - Phosphorous or Hypophosphorous Acid:
    Passes Test
  • NF - Sulfate (SO₄):
    Passes Test
  • NF - Alkali Phosphates:
    Passes Test
  • NF - Assay:
    85.0 - 88.0 %
  • EP/BP - Assay (H₃PO₄):
    84.0 - 90.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Substances Precipitated with Ammonia:
    Passes Test
  • EP/BP - Phosphorous or Hypophosphorous Acid:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤ 50 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 100 ppm
  • EP/BP - Iron (Fe):
    ≤ 50 ppm

Specification Test Results

Frequently Bought Together