Order Entry
Belgium
ContactUsLinkComponent
Lead(II) acetate trihydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Health
Environment
Lead(II) acetate trihydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Health
Environment
Lead(II) acetate trihydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    443 g/l (20 °C)
  • Melting Point:
    75 °C
  • Molar Mass:
    379.34 g/mol
  • Bulk Density:
    1200 kg/m3
  • Density:
    3.3 g/cm3 (20 °C)
  • pH:
    5.5 - 6.5 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together