Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 5 M H2SO4 (10N) volumetric solution, Fluka™
Corrosive
Sulphuric acid 5 M H2SO4 (10N) volumetric solution, Fluka™
Catalog # 35347-1L
CAS Number:  
Corrosive
Sulphuric acid 5 M H2SO4 (10N) volumetric solution, Fluka™
Catalog # 35347-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.995 - 1.005
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :98.08 g/mol
Boiling Pt:110 °C
Density:1.290 g/cm3
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589