
Sulphuric acid 2 mol/l (4 N) in aqueous solution
Catalog # VWRS5795.0250
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:98.08 g/mol
Density:1.146 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589