Order Entry
Belgium
ContactUsLinkComponent
 
Corrosive
Sulphuric acid concentrate for dilution to 2.5 mol/l (5 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4701.
Supplier:  Avantor
CAS Number:  
Corrosive
Sulphuric acid concentrate for dilution to 2.5 mol/l (5 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4701.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Titer (N):
    4.990-5.010

Specification Test Results

About this item

 :
 :98.08 g/mol
Density:1.146 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589