Order Entry
Belgium
ContactUsLinkComponent
Sodium thiosulphate standardised solution 1.0 N in aqueous solution
Sodium thiosulphate standardised solution 1.0 N in aqueous solution
Catalog # 35647.K2
CAS Number:  
Sodium thiosulphate standardised solution 1.0 N in aqueous solution
Catalog # 35647.K2
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

CAS Number:7772-98-7
 :
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
 :158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5