Order Entry
Belgium
ContactUsLinkComponent
 
Sodium thiosulphate 0.2 mol/l (0.2 N) in aqueous solution
Catalog # VWRS5722.5000
Supplier:  VWR Collection
CAS Number:  
Sodium thiosulphate 0.2 mol/l (0.2 N) in aqueous solution
Catalog # VWRS5722.5000
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499