Order Entry
Belgium
ContactUsLinkComponent
Sodium thiosulphate concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium thiosulphate concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4695.
Supplier:  Avantor
CAS Number:  
undefined
Sodium thiosulphate concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4695.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    LDPE ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501
  • Pk Info:
    50 ml

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.0998-0.1002
  • Normality (by titrimetry):
    Passes Test

Specification Test Results

About this item

 :
 :158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499