Sodium thiosulfate solution standard 0.1 mol/l (0.1 N) dans l'eau
:7772-98-7
:
:Na₂S₂O₃
:MFCD00003499
:158.11 g/mol
:Température ambiante
:231-867-5
- Appearance:Clear colorless Solution
- Titration Iodimetric:0.0950 to 0.1050 N (20°C)
- Assay:≥90 % (DAC)
- Loss on drying:≤10.0 %
- Molecular weight:600000 to 800000 Da
- Viscosity:50 to 200 mPa.s
- Ash:≤1.5 %
Frequently Bought Together