Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 4 mol/l (4 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Corrosive
Sodium hydroxide 4 mol/l (4 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # 1.11584.5000
Supplier:  Merck
CAS Number:  
Corrosive
Sodium hydroxide 4 mol/l (4 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # 1.11584.5000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Form:
    liquid
  • Amount-of-substance concentration:
    3,980-4,020
  • Measurement uncertainty:
    + / - 0,012
  • Traceability:
    NIST SRM

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,
 :M-Clarity™ quality level = MQ100
EINECS:215-185-5
UN:1824
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548