Staining trough
À propos de cet article
PMP, transparent.
Trough comes with two lids: one to help protect against evaporation when not in use; one with a handle slot facilitating the staining process.
PP tray with fold down handle can accomodate 20 slides. The close fitting lid helps protect against evaporation.
Ordering information: Trough and tray (insert) have to be ordered separately.
2 Options disponibles ci-dessous
Product Details & Documents
PMP, transparent.
Trough comes with two lids: one to help protect against evaporation when not in use; one with a handle slot facilitating the staining process.
PP tray with fold down handle can accomodate 20 slides. The close fitting lid helps protect against evaporation.
Ordering information: Trough and tray (insert) have to be ordered separately.
Product Family Options
Informations sur le produit
- W×D×HAvailabilityPrice
- BRND474300Staining trough, transparent
- BRND474305Staining trough tray, for 20 slides, 76×26 mm
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.