Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide standardised solution 2.0 N in aqueous solution
Corrosive
Sodium hydroxide standardised solution 2.0 N in aqueous solution
Catalog # 35631.K2
CAS Number:  
Corrosive
Sodium hydroxide standardised solution 2.0 N in aqueous solution
Catalog # 35631.K2
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

CAS Number:1310-73-2
 :
Formula:NaOH
MDL Number:MFCD00003548
 :40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05