ContactUsLinkComponent
Sodium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
Corrosive
Sodium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
Supplier:  Merck
Certificates
undefined
Corrosive
Sodium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
Supplier:  Merck
Certificates
Specification Test Results
  • Form:
    liquid
  • Amount-of-substance concentration:
    0.995 - 1.005
  • Measurement uncertainty:
    +/- 0.003
  • Traceability:
    NIST SRM