ContactUsLinkComponent
Sodium hydroxide standard solution 1 N in water
Corrosive
Sodium hydroxide standard solution 1 N in water
Certificates
undefined
Corrosive
Sodium hydroxide standard solution 1 N in water
Specification Test Results
  • Appearance:
    Clear colorless Solution
  • Strength:
    0.998 to 1.002 N (20°C)