Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.2 mol/l (0.2 N) in aqueous solution, AVS TITRINORM volumetric solution
Corrosive
Sodium hydroxide 0.2 mol/l (0.2 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31952.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium hydroxide 0.2 mol/l (0.2 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31952.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • NIST traceable:
    Confirmed
  • Titer (20°C; real value 0.2 % accuracy):
    0.1996 - 0.2004 mol/l

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:>105 °C (1013 hPa)
Density:1.02 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548