Staining troughs, Schiefferdecker
Staining troughs, Schiefferdecker, 70×86×51 mm
Catalog # BRND474410
Supplier: Brand
Jump to:
Product Details & Documents
Made of transparent PMP.
This rectangular jar is supplied with a lid and can hold 10 slides or 20 slides back-to-back.
Specifications
- Description:Staining troughs, Schiefferdecker
- W×D×H:70×86×51 mm