Orders LinkContactUsLinkComponent
Laboratory trays
 :   Brand
Laboratory trays
 :  Brand
Laboratory trays
Laboratory trays
PP, white.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
With ribs on base. Highly versatile trays, ideal for a wide range of applications. The ribs on the base make these trays ideal for use as developing trays.
Product Family Options
  • W×D×H
  • 225×180×45 mm
    1 * 1 ST
    1 * 1 ST - 12,90 €
    12,90 €
    1 * 1 ST
       12,90 €
  • 300×240×70 mm
    1 * 1 ST
    1 * 1 ST - 21,70 €
    21,70 €
    1 * 1 ST
       21,70 €
  • 370×310×75 mm
    1 * 1 ST
    1 * 1 ST - 31,20 €
    31,20 €
    1 * 1 ST
       31,20 €
  • 510×410×120 mm
    1 * 1 ST
    1 * 1 ST - 46,10 €
    46,10 €
    1 * 1 ST
       46,10 €
  • 652×530×140 mm
    1 * 1 ST
    1 * 1 ST - 79,70 €
    79,70 €
    1 * 1 ST
       79,70 €
  • 840×645×160 mm
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €