Order Entry
Belgium
Orders LinkContactUsLinkComponent
Staining trough, Hellendahl extended

Staining trough, Hellendahl extended

Staining trough, Hellendahl extended, 57×57×90 mm
Catalog # BRND474400
Supplier:   Brand
Staining trough, Hellendahl extended
Catalog # BRND474400
Supplier:  Brand
Staining trough, Hellendahl extended
Staining trough, Hellendahl extended

Staining trough, Hellendahl extended, 57×57×90 mm

Price
109,00 €
Log in for availability
1 * 4 ST
Product Details & Documents

Made of transparent PMP. Complete with lid.

These staining trough hold up to 16 slides (76×26 mm), 8 pairs back to back.

Specifications
  • Description:
    Staining trough, Hellendahl extended
  • W×D×H:
    57×57×90 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.