Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Supplier:   Brand
Cavity slides
Supplier:  Brand
Certificates
Cavity slides
3 Options Available Below
Product Details & Documents
Cavity slides in hydrolytic class 3 glass.
  • Ground edges
Product Family Options
Product Information
  • Colour
    No. of wells
    Well Ø
    Availability
    Price
  • BRND475505
    Cavity slides
    Cavity slides
    Clear
    1
    15 - 18 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 167,00 €
    167,00 €
    1 * 50 ST
       167,00 €
  • BRND475535
    Cavity slides
    Cavity slides
    Clear
    2
    15 - 18 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 250,00 €
    250,00 €
    1 * 50 ST
       250,00 €
  • BRND475565
    Cavity slides
    Cavity slides
    Clear
    3
    15 - 18 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 298,00 €
    298,00 €
    1 * 50 ST
       298,00 €