Orders LinkContactUsLinkComponent
Microscopy slides
Microscopy slides
Supplier:   Brand
Microscopy slides
Supplier:  Brand
Certificates
Microscopy slides
Microscopy slides
4 Options Available Below
Product Details & Documents
Microscopy slides with ground edges to reduce the risk of injury.
Product Family Options
Product Information
  • Colour
    L×W
    Availability
    Price
  • BRND474701
    Slides with cut edges
    Slides with cut edges
    White/clear
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 192,00 €
    192,00 €
    1 * 2.500 ST
       192,00 €
  • BRND474702
    Slides with cut edges and twinfrosted end
    Slides with cut edges and twinfrosted end
    White/clear
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 326,00 €
    326,00 €
    1 * 2.500 ST
       326,00 €
  • BRND474743
    Slide with ground edges
    Slide with ground edges
    White/clear
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 309,00 €
    309,00 €
    1 * 2.500 ST
       309,00 €
  • BRND474744
    Slide with ground edges, double frosted
    Slide with ground edges, double frosted
    White/clear
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 414,00 €
    414,00 €
    1 * 2.500 ST
       414,00 €