Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid concentrate for dilution to 0.05 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Sulphuric acid concentrate for dilution to 0.05 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32054.605
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sulphuric acid concentrate for dilution to 0.05 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32054.605
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    60 mL
  • Seq:
    2385
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Titer (20°C):
    0.04975 - 0.05025 mol/l

Specification Test Results

About this item

 :
 :98.08 g/mol
Boiling Pt:330
EINECS:231-639-5
UN:3264
 :GHS05
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Melting Pt:10.38
Density:1.84
ADR:8 III
 :Dose for preparation of 1 litre of solution.